Anti-IFT22 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IFT22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-255
- 77836-254
- Antigen Symbol:
- IFT22
- IFT22
- Antigen Name:
- Intraflagellar transport protein 22 homolog protein (1-185AA)
- Intraflagellar transport protein 22 homolog protein (1-185AA)
- Antigen Synonyms:
- 3110017O03Rik|AI835745|AI847427|DKFZp761N0823|FLJ13225|FLJ14117|MGC109244|RAB member of RAS oncogene family like 5|RAB member RAS oncogene family like 5|Rab-like protein 5|RABL5|RABL5_HUMAN
- 3110017O03Rik|AI835745|AI847427|DKFZp761N0823|FLJ13225|FLJ14117|MGC109244|RAB member of RAS oncogene family like 5|RAB member RAS oncogene family like 5|Rab-like protein 5|RABL5|RABL5_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H7X7
- Q9H7X7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together