Anti-SLCO1C1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLCO1C1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-597
- 77837-596
- Antigen Symbol:
- SLCO1C1
- SLCO1C1
- Antigen Name:
- Solute carrier organic anion transporter family member 1C1 protein (297-348AA)
- Solute carrier organic anion transporter family member 1C1 protein (297-348AA)
- Antigen Synonyms:
- OAT-RP-5| OATP-14| OATP-F| OATP1| OATP1C1| OATPRP5| Organic anion transporter F| Organic anion transporter polypeptide-related protein 5| organic anion transporting polypeptide 14| Organic anion-transporting polypeptide 14| SLC21A14| Slco1c1| SO1C1_HUMAN| solute carrier family 21 (organic anion transporter), member 14| Solute carrier family 21 member 14| Solute carrier organic anion transporter family member 1C1| solute carrier organic anion transporter family, member 1C1| Thyroxine transporter
- OAT-RP-5| OATP-14| OATP-F| OATP1| OATP1C1| OATPRP5| Organic anion transporter F| Organic anion transporter polypeptide-related protein 5| organic anion transporting polypeptide 14| Organic anion-transporting polypeptide 14| SLC21A14| Slco1c1| SO1C1_HUMAN| solute carrier family 21 (organic anion transporter), member 14| Solute carrier family 21 member 14| Solute carrier organic anion transporter family member 1C1| solute carrier organic anion transporter family, member 1C1| Thyroxine transporter
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NYB5
- Q9NYB5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together