Anti-UNC50 Rabbit Recombinant Polyclonal Antibody
77826-981
New Product
- Antigen Name:Protein unc-50 homolog protein (15-82AA)
- Antigen Symbol:UNC50
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q53HI1
- Antigen Synonyms:UNC50|UNCL|HSD-23|HSD23|Protein unc-50 homolog|Periodontal ligament-specific protein 22|PDLs22|Protein GMH1 homolog|hGMH1|Uncoordinated-like protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000, IF: 1:50 to 1:200
Type:
Antigen: UNC50
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human