Anti-CCS Rabbit Polyclonal Antibody
77814-379
New Product
- Antigen Name:Synthesized peptide derived from C-terminal of CCS
- Antigen Symbol:CCS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O14618
- Antigen Synonyms:CCS_HUMAN|Copper chaperone for superoxide dismutase|MGC138260|SOD 4|SOD4|Superoxide dismutase copper chaperone
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: CCS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human