Anti-SHC1 Rabbit Polyclonal Antibody
Catalog # 77808-079
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from Shc around the non-phosphorylation site of Y349.
- Antigen Symbol:SHC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 μg
- Form:Liquid
- Gene ID:P29353
- Antigen Synonyms:FLJ26504|p66|p66SHC|SH2 domain protein C1|SHC (Src homology 2 domain containing) transforming protein 1|SHC 1|SHC A|SHC adaptor protein 1|Shc|SHC transforming protein 1|SHC transforming protein|SHC-transforming protein 1|SHC-transforming protein 3|SHC-transforming protein A|SHC1_HUMAN|SHCA|Src homology 2 domain-containing-transforming protein C1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, IP: 2 to 5 μg, ELISA: 1:20000
Type:
Antigen: SHC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat