Anti-TRABD2B Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRABD2B
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77818-098
- 77818-099
- Antigen Symbol:
- TRABD2B
- TRABD2B
- Antigen Name:
- Metalloprotease TIKI2 protein (195-405AA)
- Metalloprotease TIKI2 protein (195-405AA)
- Antigen Synonyms:
- TRABD2B|HKAT|TIKI2|Metalloprotease TIKI2|EC 3.4.-.-|Heart|kidney and adipose-enriched transmembrane protein homolog|TRAB domain-containing protein 2B
- TRABD2B|HKAT|TIKI2|Metalloprotease TIKI2|EC 3.4.-.-|Heart|kidney and adipose-enriched transmembrane protein homolog|TRAB domain-containing protein 2B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A6NFA1
- A6NFA1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together