Anti-WAPL Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: WAPL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-302
- 77832-303
- Antigen Symbol:
- WAPL
- WAPL
- Antigen Name:
- Wings apart-like protein homolog protein (1037-1190AA)
- Wings apart-like protein homolog protein (1037-1190AA)
- Antigen Synonyms:
- FOE| Friend of EBNA2 (Epstein Barr virus nuclear protein 2)| Friend of EBNA2 protein| KIAA0261| OTTHUMP00000020004| WAPAL| WAPL| WAPL_HUMAN| Wings apart like homolog| Wings apart like homolog (Drosophila)| Wings apart-like protein homolog
- FOE| Friend of EBNA2 (Epstein Barr virus nuclear protein 2)| Friend of EBNA2 protein| KIAA0261| OTTHUMP00000020004| WAPAL| WAPL| WAPL_HUMAN| Wings apart like homolog| Wings apart like homolog (Drosophila)| Wings apart-like protein homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z5K2
- Q7Z5K2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together