Anti-CFLAR Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: CFLAR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
- Catalog No:
- 77815-744
- 77815-743
- Antigen Symbol:
- CFLAR
- CFLAR
- Antigen Name:
- Synthetic peptide of CFLAR
- Synthetic peptide of CFLAR
- Antigen Synonyms:
- c FLIP| c FLIPL| c FLIPR| c FLIPS| c-FLIP| c-FLIPL| c-FLIPR| c-FLIPS| CASH| CASP8 and FADD like apoptosis regulator| CASP8 and FADD-like apoptosis regulator subunit p12| CASP8AP1| Caspase homolog| Caspase like apoptosis regulatory protein| Caspase related inducer of apoptosis| Caspase-eight-related protein| Caspase-like apoptosis regulatory protein| Casper| cellular flice inhibitory protein| Cellular FLICE-like inhibitory protein| CFLA| Cflar| CFLAR_HUMAN| CLARP| FADD like anti apoptotic molecule| FADD-like antiapoptotic molecule 1| FLAME| FLAME-1| FLAME1| FLIP| I FLICE| I-FLICE| Inhibitor of FLICE| MACH-related inducer of toxicity| MRIT| OTTHUMP00000163715| OTTHUMP00000206475| OTTHUMP00000206476| OTTHUMP00000206478| OTTHUMP00000206479| OTTHUMP00000206480| OTTHUMP00000206482| OTTHUMP00000207360| Usurpin| Usurpin beta
- c FLIP| c FLIPL| c FLIPR| c FLIPS| c-FLIP| c-FLIPL| c-FLIPR| c-FLIPS| CASH| CASP8 and FADD like apoptosis regulator| CASP8 and FADD-like apoptosis regulator subunit p12| CASP8AP1| Caspase homolog| Caspase like apoptosis regulatory protein| Caspase related inducer of apoptosis| Caspase-eight-related protein| Caspase-like apoptosis regulatory protein| Casper| cellular flice inhibitory protein| Cellular FLICE-like inhibitory protein| CFLA| Cflar| CFLAR_HUMAN| CLARP| FADD like anti apoptotic molecule| FADD-like antiapoptotic molecule 1| FLAME| FLAME-1| FLAME1| FLIP| I FLICE| I-FLICE| Inhibitor of FLICE| MACH-related inducer of toxicity| MRIT| OTTHUMP00000163715| OTTHUMP00000206475| OTTHUMP00000206476| OTTHUMP00000206478| OTTHUMP00000206479| OTTHUMP00000206480| OTTHUMP00000206482| OTTHUMP00000207360| Usurpin| Usurpin beta
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15519
- O15519
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together