Anti-MRPL28 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:200 to 1:1000
Type:
Antigen: MRPL28
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-283
- 77824-282
- Antigen Symbol:
- MRPL28
- MRPL28
- Antigen Name:
- 39S ribosomal protein L28, mitochondrial protein (1-256AA)
- 39S ribosomal protein L28, mitochondrial protein (1-256AA)
- Antigen Synonyms:
- 39S ribosomal protein L28|39S ribosomal protein L28 mitochondrial|39S ribosomal protein L28 mitochondrial precursor|HGNC6756|L28mt|MAAT 1|MAAT1|Melanoma antigen p15|Melanoma associated antigen recognised by cytotoxic T lymphocytes|Melanoma associated antigen recognized by T lymphocytes|Melanoma-associated antigen recognized by T-lymphocytes|MGC8499|mitochondrial|Mitochondrial ribosomal protein L28|MRP L28|MRP-L28|MRPL 28|mrpl28|P15|RM28_HUMAN
- 39S ribosomal protein L28|39S ribosomal protein L28 mitochondrial|39S ribosomal protein L28 mitochondrial precursor|HGNC6756|L28mt|MAAT 1|MAAT1|Melanoma antigen p15|Melanoma associated antigen recognised by cytotoxic T lymphocytes|Melanoma associated antigen recognized by T lymphocytes|Melanoma-associated antigen recognized by T-lymphocytes|MGC8499|mitochondrial|Mitochondrial ribosomal protein L28|MRP L28|MRP-L28|MRPL 28|mrpl28|P15|RM28_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13084
- Q13084
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together