Anti-RC3H2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RC3H2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-165
- 77842-166
- Antigen Symbol:
- RC3H2
- RC3H2
- Antigen Name:
- Roquin-2 protein (251-506AA)
- Roquin-2 protein (251-506AA)
- Antigen Synonyms:
- 2900024N03Rik| 9430019J22Rik| D930043C02Rik| FLJ20301| FLJ20713| Membrane associated DNA binding protein| Membrane associated nucleic acid binding protein| Membrane-associated nucleic acid-binding protein| MGC52176| MNAB| OTTHUMP00000022076| OTTHUMP00000022077| OTTMUSP00000013545| OTTMUSP00000013546| Rc3h2| RC3H2_HUMAN| Ring finger and CCCH type domains 2| RING finger and CCCH type zinc finger domain containing protein 2| Ring finger and CCCH-type zinc finger domain-containing protein 2| ring finger and CCCH-type zinc finger domains 2| RING finger protein 164| RNF164| roquin-2| RP23-321B3.1
- 2900024N03Rik| 9430019J22Rik| D930043C02Rik| FLJ20301| FLJ20713| Membrane associated DNA binding protein| Membrane associated nucleic acid binding protein| Membrane-associated nucleic acid-binding protein| MGC52176| MNAB| OTTHUMP00000022076| OTTHUMP00000022077| OTTMUSP00000013545| OTTMUSP00000013546| Rc3h2| RC3H2_HUMAN| Ring finger and CCCH type domains 2| RING finger and CCCH type zinc finger domain containing protein 2| Ring finger and CCCH-type zinc finger domain-containing protein 2| ring finger and CCCH-type zinc finger domains 2| RING finger protein 164| RNF164| roquin-2| RP23-321B3.1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HBD1
- Q9HBD1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together