Anti-CATSPERE Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: CATSPERE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77828-183
- 77828-184
- Antigen Symbol:
- CATSPERE
- CATSPERE
- Antigen Name:
- Cation channel sperm-associated protein subunit epsilon protein (601-832AA)
- Cation channel sperm-associated protein subunit epsilon protein (601-832AA)
- Antigen Synonyms:
- C1orf101|CA101_HUMAN|Chromosome 1 Open Reading Frame 101|RP11-523K4.1|Uncharacterized protein C1orf101
- C1orf101|CA101_HUMAN|Chromosome 1 Open Reading Frame 101|RP11-523K4.1|Uncharacterized protein C1orf101
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5SY80
- Q5SY80
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together