Anti-CWC22 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: CWC22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77839-555
- 77839-556
- Antigen Symbol:
- CWC22
- CWC22
- Antigen Name:
- Pre-mRNA-splicing factor CWC22 homolog protein (360-623AA)
- Pre-mRNA-splicing factor CWC22 homolog protein (360-623AA)
- Antigen Synonyms:
- CWC22 spliceosome associated protein homolog (S. cerevisiae)|CWC22 spliceosome associated protein homolog|CWC22_HUMAN|EIF4GL|fSAPb|Functional spliceosome associated protein b|KIAA1604|NCM|Nucampholin homolog|Pre-mRNA-splicing factor CWC22 homolog
- CWC22 spliceosome associated protein homolog (S. cerevisiae)|CWC22 spliceosome associated protein homolog|CWC22_HUMAN|EIF4GL|fSAPb|Functional spliceosome associated protein b|KIAA1604|NCM|Nucampholin homolog|Pre-mRNA-splicing factor CWC22 homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HCG8
- Q9HCG8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together