Anti-KPNB1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77824-912
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Importin subunit beta-1 protein (9-214AA)
- Antigen Symbol:KPNB1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q14974
- Antigen Synonyms:IMB 1|IMB1|IMB1_HUMAN|Impnb|Importin 90|Importin beta 1|Importin beta 1 subunit|Importin subunit beta-1|Importin-90|IPOB|Karyopherin beta 1|Karyopherin beta 1 subunit|Karyopherin subunit beta-1|KPNB 1|Kpnb1|MGC2155|MGC2156|MGC2157|NTF 97|NTF97|NTF97/Importin beta|Nuclear factor P97|Pore targeting complex 97 kDa subunit|PTAC97
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:200 to 1:500
Type:
Antigen: KPNB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human