Anti-JUND Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: JUND
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77807-744
- 77807-743
- Antigen Symbol:
- JUND
- JUND
- Antigen Name:
- Transcription factor jun-D protein (1-136AA)
- Transcription factor jun-D protein (1-136AA)
- Antigen Synonyms:
- Activator protein 1|AP 1|AP1|Jun D|jun D proto oncogene|JunD FL isoform|JUND_HUMAN|Transcription factor jun D|Transcription factor jun-D
- Activator protein 1|AP 1|AP1|Jun D|jun D proto oncogene|JunD FL isoform|JUND_HUMAN|Transcription factor jun D|Transcription factor jun-D
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17535
- P17535
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together