Anti-E2F5 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: E2F5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77825-447
- 77825-448
- Antigen Symbol:
- E2F5
- E2F5
- Antigen Name:
- Transcription factor E2F5 protein (193-298AA)
- Transcription factor E2F5 protein (193-298AA)
- Antigen Synonyms:
- E2F 5| E2F transcription factor 5| E2F transcription factor 5 p130 binding| E2F-5| E2f5| E2F5_HUMAN| p130 binding| Transcription factor E2F5
- E2F 5| E2F transcription factor 5| E2F transcription factor 5 p130 binding| E2F-5| E2f5| E2F5_HUMAN| p130 binding| Transcription factor E2F5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15329
- Q15329
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together