Anti-RND2 Rabbit Polyclonal Antibody
77797-420
New Product
- Antigen Name:RND2_HUMAN
- Antigen Symbol:RND2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P52198
- Antigen Synonyms:ARHN|CTD-3199J23.4|GTP-binding protein Rho7|ras homolog gene family, member N|Rho 7|Rho family GTPase 2|Rho N|Rho related GTP-binding protein RhoN|Rho-related GTP-binding protein Rho7|Rho-related GTP-binding protein RhoN|Rho7|RhoN|Rnd 2|Rnd2|RND2_HUMAN
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: RND2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat