Anti-PAEP Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:100 to 1:300
Type:
Antigen: PAEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse
Specifications
- Catalog No:
- 77807-687
- 77807-688
- Antigen Symbol:
- PAEP
- PAEP
- Antigen Name:
- Fusion protein of PAEP
- Fusion protein of PAEP
- Antigen Synonyms:
- Alpha uterine protein|gD|GdA|GdF|GdS|Glycodelin A|Glycodelin|Glycodelin F|Glycodelin S|MGC138509|MGC142288|PAEG|PAEP|PAEP_HUMAN|PEG|PEP|Placental Protein 14 / Glycodelin A|Placental protein 14|PP14|Pregnancy-associated endometrial alpha-2 globulin|Progestagen-associated endometrial protein|Progesterone-associated endometrial protein
- Alpha uterine protein|gD|GdA|GdF|GdS|Glycodelin A|Glycodelin|Glycodelin F|Glycodelin S|MGC138509|MGC142288|PAEG|PAEP|PAEP_HUMAN|PEG|PEP|Placental Protein 14 / Glycodelin A|Placental protein 14|PP14|Pregnancy-associated endometrial alpha-2 globulin|Progestagen-associated endometrial protein|Progesterone-associated endometrial protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P09466
- P09466
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together