Anti-WWOX Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: WWOX
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-918
- 77838-917
- Antigen Symbol:
- WWOX
- WWOX
- Antigen Name:
- WW domain-containing oxidoreductase protein (1-180AA)
- WW domain-containing oxidoreductase protein (1-180AA)
- Antigen Synonyms:
- 5330426P09Rik|9030416C10Rik|Aberrant WW domain-containing oxidoreductase|D16S432E|EC 1.1.1.-|EIEE28|FOR|FRA16D|Fragile site FRA16D oxidoreductase|Fragile site FRA16D Oxireductase|HHCMA56|MGC55975|PRO0128|Putative oxidoreductase|SCAR12|SDR41C1|Short chain dehydrogenase/reductase family 41C, member 1|WOX1|WW domain containing oxidoreductase|WW domain-containing oxidoreductase|WW domain-containing protein WWOX|wwox|WWOX_HUMAN|zgc:55975
- 5330426P09Rik|9030416C10Rik|Aberrant WW domain-containing oxidoreductase|D16S432E|EC 1.1.1.-|EIEE28|FOR|FRA16D|Fragile site FRA16D oxidoreductase|Fragile site FRA16D Oxireductase|HHCMA56|MGC55975|PRO0128|Putative oxidoreductase|SCAR12|SDR41C1|Short chain dehydrogenase/reductase family 41C, member 1|WOX1|WW domain containing oxidoreductase|WW domain-containing oxidoreductase|WW domain-containing protein WWOX|wwox|WWOX_HUMAN|zgc:55975
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZC7
- Q9NZC7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together