Anti-LDLRAP1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: LDLRAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-738
- 77828-739
- Antigen Symbol:
- LDLRAP1
- LDLRAP1
- Antigen Name:
- Low density lipoprotein receptor adapter protein 1 protein (59-308AA)
- Low density lipoprotein receptor adapter protein 1 protein (59-308AA)
- Antigen Synonyms:
- ARH|ARH GENE|ARH_HUMAN|ARH1|ARH2|Autosomal recessive hypercholesterolemia protein|DKFZp586D0624|FHCB1|FHCB2|LDL receptor adaptor protein|Ldlrap1|Low density lipoprotein receptor adapter protein 1|MGC34705|OTTHUMP00000008526
- ARH|ARH GENE|ARH_HUMAN|ARH1|ARH2|Autosomal recessive hypercholesterolemia protein|DKFZp586D0624|FHCB1|FHCB2|LDL receptor adaptor protein|Ldlrap1|Low density lipoprotein receptor adapter protein 1|MGC34705|OTTHUMP00000008526
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5SW96
- Q5SW96
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together