Anti-TTC33 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TTC33
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77830-071
- 77830-072
- Antigen Symbol:
- TTC33
- TTC33
- Antigen Name:
- Tetratricopeptide repeat protein 33 protein (1-262AA)
- Tetratricopeptide repeat protein 33 protein (1-262AA)
- Antigen Synonyms:
- Osmosis responsive factor|Osmosis-responsive factor|OSRF|Tetratricopeptide repeat domain 33|Tetratricopeptide repeat protein 33|TPR repeat protein 33|TTC33|TTC33_HUMAN
- Osmosis responsive factor|Osmosis-responsive factor|OSRF|Tetratricopeptide repeat domain 33|Tetratricopeptide repeat protein 33|TPR repeat protein 33|TTC33|TTC33_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6PID6
- Q6PID6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together