Anti-ZNF695 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZNF695
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77805-798
- 77805-799
- Antigen Symbol:
- ZNF695
- ZNF695
- Antigen Name:
- Zinc finger protein 695 protein (1-88AA)
- Zinc finger protein 695 protein (1-88AA)
- Antigen Synonyms:
- MGC61907 |RP11-551G24.1|SBZF3|Zinc finger protein 695|Zinc finger protein SBZF3|ZN695_HUMAN|ZNF695
- MGC61907 |RP11-551G24.1|SBZF3|Zinc finger protein 695|Zinc finger protein SBZF3|ZN695_HUMAN|ZNF695
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IW36
- Q8IW36
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together