Order Entry
United States
ContactUsLinkComponent
Anti-TADA3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-TADA3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-TADA3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77806-811
  • 77806-810
  • Antigen Symbol:
  • TADA3
  • TADA3
  • Antigen Name:
  • Transcriptional adapter 3 protein (1-238AA)
  • Transcriptional adapter 3 protein (1-238AA)
  • Antigen Synonyms:
  • TADA3| ADA3| TADA3L| Transcriptional adapter 3| ADA3 homolog| hADA3| STAF54| Transcriptional adapter 3-like| ADA3-like protein
  • TADA3| ADA3| TADA3L| Transcriptional adapter 3| ADA3 homolog| hADA3| STAF54| Transcriptional adapter 3-like| ADA3-like protein
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O75528
  • O75528
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together