Anti-POT1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: POT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-493
- 77837-492
- Antigen Symbol:
- POT1
- POT1
- Antigen Name:
- Protection of telomeres protein 1 protein (250-432AA)
- Protection of telomeres protein 1 protein (250-432AA)
- Antigen Synonyms:
- DKFZP586D211|hPot 1|hPot1|POT 1|POT1|POT1 like telomere end-binding protein|POT1 protection of telomeres 1 homolog|POT1-like telomere end-binding protein|POTE1_HUMAN|Protection of telomeres 1|Protection of telomeres 1 homolog (S. pombe)|Protection of telomeres protein 1
- DKFZP586D211|hPot 1|hPot1|POT 1|POT1|POT1 like telomere end-binding protein|POT1 protection of telomeres 1 homolog|POT1-like telomere end-binding protein|POTE1_HUMAN|Protection of telomeres 1|Protection of telomeres 1 homolog (S. pombe)|Protection of telomeres protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NUX5
- Q9NUX5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together