Anti-GAS2L1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: GAS2L1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-817
- 77834-816
- Antigen Symbol:
- GAS2L1
- GAS2L1
- Antigen Name:
- GAS2-like protein 1 protein (130-400AA)
- GAS2-like protein 1 protein (130-400AA)
- Antigen Synonyms:
- GA2L1_HUMAN|GAR 22|GAR22|GAS2 like protein 1|GAS2 related protein on chromosome 22|GAS2-like protein 1|GAS2-related protein on chromosome 22|GAS2L1|Growth arrest specific 2 like 1|Growth arrest specific protein 2 like 1|Growth arrest-specific protein 2-like 1|MGC17243
- GA2L1_HUMAN|GAR 22|GAR22|GAS2 like protein 1|GAS2 related protein on chromosome 22|GAS2-like protein 1|GAS2-related protein on chromosome 22|GAS2L1|Growth arrest specific 2 like 1|Growth arrest specific protein 2 like 1|Growth arrest-specific protein 2-like 1|MGC17243
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99501
- Q99501
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together