Anti-UGT2B7 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UGT2B7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-983
- 77803-983
- Antigen Symbol:
- UGT2B7
- UGT2B7
- Antigen Name:
- UDP-glucuronosyltransferase 2B7 protein (237-382AA)
- UDP-glucuronosyltransferase 2B7 protein (237-382AA)
- Antigen Synonyms:
- UDP glucuronosyltransferase 2 family, member B7|3 4 catechol estrogen specific|3|3,4 catechol estrogen specific UDPGT|4-catechol estrogen-specific UDPGT|UD2B7_HUMAN|UDP glucuronosyltransferase 2 family, polypeptide B7|UDP glucuronosyltransferase 2 family polypeptide B7|UDP glucuronosyltransferase 2 family, member B9|UDP glucuronosyltransferase 2B7|UDP glucuronosyltransferase2 family, polypeptide B7|UDP glucuronyltransferase family 2 beta 7|UDP glycosyltransferase 2 family polypeptide B7|UDP-glucuronosyltransferase 2B7|UDP-glucuronosyltransferase 2B9|UDPGT|UDPGT 2B7|UDPGT 2B9|UDPGT2B7|UDPGTh 2|UDPGTh-2|UDPGTh2|UGT2B7|UGT2B9|UGTB2B9|uridine diphosphate glycosyltransferase 2 family, member B7|uridine diphosphate glycosyltransferase 2 family, member B9
- UDP glucuronosyltransferase 2 family, member B7|3 4 catechol estrogen specific|3|3,4 catechol estrogen specific UDPGT|4-catechol estrogen-specific UDPGT|UD2B7_HUMAN|UDP glucuronosyltransferase 2 family, polypeptide B7|UDP glucuronosyltransferase 2 family polypeptide B7|UDP glucuronosyltransferase 2 family, member B9|UDP glucuronosyltransferase 2B7|UDP glucuronosyltransferase2 family, polypeptide B7|UDP glucuronyltransferase family 2 beta 7|UDP glycosyltransferase 2 family polypeptide B7|UDP-glucuronosyltransferase 2B7|UDP-glucuronosyltransferase 2B9|UDPGT|UDPGT 2B7|UDPGT 2B9|UDPGT2B7|UDPGTh 2|UDPGTh-2|UDPGTh2|UGT2B7|UGT2B9|UGTB2B9|uridine diphosphate glycosyltransferase 2 family, member B7|uridine diphosphate glycosyltransferase 2 family, member B9
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P16662
- P16662
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together