Anti-IFT27 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IFT27
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-165
- 77839-164
- Antigen Symbol:
- IFT27
- IFT27
- Antigen Name:
- Intraflagellar transport protein 27 homolog protein (1-186AA)
- Intraflagellar transport protein 27 homolog protein (1-186AA)
- Antigen Synonyms:
- BBS19|Ift27|IFT27_HUMAN|Intraflagellar transport 27|Intraflagellar transport protein 27 homolog|Putative GTP binding protein RAY like|Putative GTP-binding protein RAY-like|Rab like protein 4|RAB member of RAS oncogene family like 4|Rab-like protein 4|RAYL
- BBS19|Ift27|IFT27_HUMAN|Intraflagellar transport 27|Intraflagellar transport protein 27 homolog|Putative GTP binding protein RAY like|Putative GTP-binding protein RAY-like|Rab like protein 4|RAB member of RAS oncogene family like 4|Rab-like protein 4|RAYL
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BW83
- Q9BW83
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together