Anti-TSC22D1 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: TSC22D1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77825-026
- 77825-027
- Antigen Symbol:
- TSC22D1
- TSC22D1
- Antigen Name:
- TSC22 domain family protein 1 protein (1-144AA)
- TSC22 domain family protein 1 protein (1-144AA)
- Antigen Synonyms:
- Cerebral protein 2|DKFZp686O19206|Hucep 2|Hucep2|KIAA1994|MGC17597|OTTHUMP00000040978|Ptg 2|Regulatory protein TSC 22|Regulatory protein TSC-22|Regulatory protein TSC22|RP11 269C23.2|T22D1_HUMAN|TGFB stimulated clone 22 homolog|TGFB-stimulated clone 22 homolog|TGFB1I4|TGFbeta stimulated clone 22|Transcriptional regulator TSC 22|Transforming growth factor beta 1 induced transcript 4|Transforming growth factor beta 1 induced transcript 4 protein|Transforming growth factor beta stimulated protein TSC 22|Transforming growth factor beta-1-induced transcript 4 protein|TSC 22|TSC22|TSC22 domain family 1 isoform 2|TSC22 domain family member 1|TSC22 domain family protein 1|TSC22D1
- Cerebral protein 2|DKFZp686O19206|Hucep 2|Hucep2|KIAA1994|MGC17597|OTTHUMP00000040978|Ptg 2|Regulatory protein TSC 22|Regulatory protein TSC-22|Regulatory protein TSC22|RP11 269C23.2|T22D1_HUMAN|TGFB stimulated clone 22 homolog|TGFB-stimulated clone 22 homolog|TGFB1I4|TGFbeta stimulated clone 22|Transcriptional regulator TSC 22|Transforming growth factor beta 1 induced transcript 4|Transforming growth factor beta 1 induced transcript 4 protein|Transforming growth factor beta stimulated protein TSC 22|Transforming growth factor beta-1-induced transcript 4 protein|TSC 22|TSC22|TSC22 domain family 1 isoform 2|TSC22 domain family member 1|TSC22 domain family protein 1|TSC22D1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15714
- Q15714
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together