Anti-SNRK Rabbit Polyclonal Antibody
Catalog # 77837-424
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:SNF-related serine/threonine-protein kinase protein (340-360AA)
- Antigen Symbol:SNRK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9NRH2
- Antigen Synonyms:DKFZp779A1866|FLJ20224|HSNFRK|KIAA0096|SNF related kinase|SNF related serine/threonine protein kinase|SNF-related serine/threonine-protein kinase|SNF1 related kinase|SNF1-related kinase|SNFRK|Snrk|SNRK_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: SNRK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human