Anti-IFI27L2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IFI27L2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-990
- 77841-991
- Antigen Symbol:
- IFI27L2
- IFI27L2
- Antigen Name:
- Interferon alpha-inducible protein 27-like protein 2 protein (87-130AA)
- Interferon alpha-inducible protein 27-like protein 2 protein (87-130AA)
- Antigen Synonyms:
- IFI27L2| FAM14A| TLH29| Interferon alpha-inducible protein 27-like protein 2| Interferon-stimulated gene 12b protein| ISG12(b)| ISG12B| Protein TLH29| pIFI27-like protein
- IFI27L2| FAM14A| TLH29| Interferon alpha-inducible protein 27-like protein 2| Interferon-stimulated gene 12b protein| ISG12(b)| ISG12B| Protein TLH29| pIFI27-like protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H2X8
- Q9H2X8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together