Anti-OLFM4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: OLFM4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-489
- 77831-490
- Antigen Symbol:
- OLFM4
- OLFM4
- Antigen Name:
- Olfactomedin-4 protein (224-334AA)
- Olfactomedin-4 protein (224-334AA)
- Antigen Synonyms:
- Antiapoptotic protein GW112|bA209J19.1|G-CSF-stimulated clone 1 protein|GC1|GW112|hGC 1|hGC-1|hOLfD|KIAA4294|olfactomedin 4|Olfactomedin-4|OlfD|OLFM4|OLFM4_HUMAN|OLM4|UNQ362
- Antiapoptotic protein GW112|bA209J19.1|G-CSF-stimulated clone 1 protein|GC1|GW112|hGC 1|hGC-1|hOLfD|KIAA4294|olfactomedin 4|Olfactomedin-4|OlfD|OLFM4|OLFM4_HUMAN|OLM4|UNQ362
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6UX06
- Q6UX06
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together