Anti-Ripk1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Ripk1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77828-291
- 77828-292
- Antigen Symbol:
- Ripk1
- Ripk1
- Antigen Name:
- Recombinant Mouse Receptor-interacting serine/threonine-protein kinase 1 protein (1-656AA)
- Recombinant Mouse Receptor-interacting serine/threonine-protein kinase 1 protein (1-656AA)
- Antigen Synonyms:
- Ripk1|Rinp|Rip|Receptor-interacting serine/threonine-protein kinase 1|EC 2.7.11.1|Cell death protein RIP|Receptor-interacting protein 1|RIP-1
- Ripk1|Rinp|Rip|Receptor-interacting serine/threonine-protein kinase 1|EC 2.7.11.1|Cell death protein RIP|Receptor-interacting protein 1|RIP-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q60855
- Q60855
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together