Anti-DCAF4 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DCAF4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-635
- 77833-634
- Antigen Symbol:
- DCAF4
- DCAF4
- Antigen Name:
- DDB1- and CUL4-associated factor 4 protein (1-495AA)
- DDB1- and CUL4-associated factor 4 protein (1-495AA)
- Antigen Synonyms:
- DCAF4|DCAF4_HUMAN|DDB1 and CUL4 associated factor 4|DDB1- and CUL4-associated factor 4|WD repeat domain 21|WD repeat domain 21A|WD repeat-containing protein 21A|WDR21|WDR21A
- DCAF4|DCAF4_HUMAN|DDB1 and CUL4 associated factor 4|DDB1- and CUL4-associated factor 4|WD repeat domain 21|WD repeat domain 21A|WD repeat-containing protein 21A|WDR21|WDR21A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WV16
- Q8WV16
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together