Anti-STRADB Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: STRADB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-273
- 77839-272
- Antigen Symbol:
- STRADB
- STRADB
- Antigen Name:
- STE20-related kinase adapter protein beta protein (212-418AA)
- STE20-related kinase adapter protein beta protein (212-418AA)
- Antigen Synonyms:
- ALS2 chromosome region gene 2|ALS2CR2|Amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 2|Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein|CALS 21|CALS-21|ILP interacting protein ILPIPA|ILP-interacting protein|ILPIP|ILPIPA|Likely ortholog of mouse polyploidy associated protein kinase|PAPK|PRO1038|Pseudokinase ALS2CR2|STE20-related kinase adapter protein beta|STRAB_HUMAN|STRAD beta|STRADB
- ALS2 chromosome region gene 2|ALS2CR2|Amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 2|Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein|CALS 21|CALS-21|ILP interacting protein ILPIPA|ILP-interacting protein|ILPIP|ILPIPA|Likely ortholog of mouse polyploidy associated protein kinase|PAPK|PRO1038|Pseudokinase ALS2CR2|STE20-related kinase adapter protein beta|STRAB_HUMAN|STRAD beta|STRADB
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9C0K7
- Q9C0K7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together