Anti-TAC1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:20 to 1:100
Type:
Antigen: TAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77821-175
- 77821-176
- Antigen Symbol:
- TAC1
- TAC1
- Antigen Name:
- Fusion protein of TAC1
- Fusion protein of TAC1
- Antigen Synonyms:
- C-terminal-flanking peptide|Hs.2563|neurokinin 1|neurokinin 2|neurokinin A|neurokinin alpha|Neuromedin L|neuropeptide gamma|neuropeptide K|NK2|NKA|NKNA|NPK|PPT|preprotachykinin|protachykinin|protachykinin-1|Substance K|SubstanceP|TAC1|TAC2|formerly|Tachykinin 1|tachykinin 2|Tachykinin precursor 1|tachykinin|precursor 1 (substance K|substance P|neuropeptide gamma)|Tachykinin1|TKN1_HUMAN
- C-terminal-flanking peptide|Hs.2563|neurokinin 1|neurokinin 2|neurokinin A|neurokinin alpha|Neuromedin L|neuropeptide gamma|neuropeptide K|NK2|NKA|NKNA|NPK|PPT|preprotachykinin|protachykinin|protachykinin-1|Substance K|SubstanceP|TAC1|TAC2|formerly|Tachykinin 1|tachykinin 2|Tachykinin precursor 1|tachykinin|precursor 1 (substance K|substance P|neuropeptide gamma)|Tachykinin1|TKN1_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20366
- P20366
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together