Anti-KCNIP1 Rabbit Polyclonal Antibody
Catalog # 77821-027
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthetic Peptide
- Antigen Symbol:KCNIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NZI2
- Antigen Synonyms:A type potassium channel modulatory protein 1|A-type potassium channel modulatory protein 1|KChIP1|KCIP1_HUMAN|Kcnip1|Kv channel interacting protein 1|Kv channel-interacting protein 1|MGC9|MGC95|Potassium channel interacting protein 1|Potassium channel-interacting protein 1|VABP|Vesicle APC binding protein|Vesicle APC-binding protein
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:100 to 200
Type:
Antigen: KCNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse