Anti-SPACA3 Rabbit Polyclonal Antibody
77800-780
New Product
- Antigen Name:processed form]
- Antigen Symbol:SPACA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8IXA5
- Antigen Synonyms:SPACA3|LYC3|LYZL3|SLLP1|SPRASA|UNQ424/PRO862Sperm acrosome membrane-associated protein 3|Cancer/testis antigen 54|CT54|Lysozyme-like acrosomal sperm-specific secretory protein ALLP-17|Lysozyme-like protein 3|Sperm lysozyme-like protein 1|Sperm protein reactive with antisperm antibodies|Sperm protein reactive with ASA) [Cleaved into: Sperm acrosome membrane-associated protein 3|membrane form|Sperm acrosome membrane-associated protein 3|processed form]
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: SPACA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat