Anti-BRD7 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: BRD7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-395
- 77836-396
- Antigen Symbol:
- BRD7
- BRD7
- Antigen Name:
- Bromodomain-containing protein 7 protein (601-651AA)
- Bromodomain-containing protein 7 protein (601-651AA)
- Antigen Synonyms:
- 75 kDa bromodomain protein|BP75|BRD 7|BRD7_HUMAN|Bromodomain containing 7|bromodomain containing protein 7|Bromodomain-containing protein 7|CELTIX 1|CELTIX1|NAG4|Protein CELTIX-1
- 75 kDa bromodomain protein|BP75|BRD 7|BRD7_HUMAN|Bromodomain containing 7|bromodomain containing protein 7|Bromodomain-containing protein 7|CELTIX 1|CELTIX1|NAG4|Protein CELTIX-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NPI1
- Q9NPI1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together