Anti-COPS7A Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: COPS7A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-183
- 77840-184
- Antigen Symbol:
- COPS7A
- COPS7A
- Antigen Name:
- COP9 signalosome complex subunit 7a protein (2-275AA)
- COP9 signalosome complex subunit 7a protein (2-275AA)
- Antigen Synonyms:
- COP9 complex subunit 7a| COP9 constitutive photomorphogenic homolog subunit 7A| COP9 signalosome complex subunit 7a| COP9 signalosome subunit 7A| COPS7A| CSN7A| CSN7A_HUMAN| Dermal papilla derived protein 10| Dermal papilla-derived protein 10| DERP10| JAB1 containing signalosome subunit 7a| JAB1-containing signalosome subunit 7a| SGN7a| Signalosome subunit 7a
- COP9 complex subunit 7a| COP9 constitutive photomorphogenic homolog subunit 7A| COP9 signalosome complex subunit 7a| COP9 signalosome subunit 7A| COPS7A| CSN7A| CSN7A_HUMAN| Dermal papilla derived protein 10| Dermal papilla-derived protein 10| DERP10| JAB1 containing signalosome subunit 7a| JAB1-containing signalosome subunit 7a| SGN7a| Signalosome subunit 7a
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UBW8
- Q9UBW8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together