Anti-PDE1C Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PDE1C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-288
- 77825-287
- Antigen Symbol:
- PDE1C
- PDE1C
- Antigen Name:
- Calcium/calmodulin-dependent 3\',5\'-cyclic nucleotide phosphodiesterase 1C protein (542-634AA)
- Calcium/calmodulin-dependent 3\',5\'-cyclic nucleotide phosphodiesterase 1C protein (542-634AA)
- Antigen Synonyms:
- 5''-cyclic nucleotide phosphodiesterase 1C|Calcium/calmodulin dependent 3'5' cyclic nucleotide phosphodiesterase 1C|Calcium/calmodulin-dependent 3''|Cam PDE 1C|Cam-PDE 1C|hCam 3|hCam-3|HCAM3|HSPDE1C1A|Human 3'5' cyclic nucleotide Phosphodiesterase|PDE 1C|Pde1c|PDE1C_HUMAN|Phosphodiesterase 1C|Phosphodiesterase 1C calmodulin dependent|Phosphodiesterase 1C calmodulin dependent 70kDa
- 5''-cyclic nucleotide phosphodiesterase 1C|Calcium/calmodulin dependent 3'5' cyclic nucleotide phosphodiesterase 1C|Calcium/calmodulin-dependent 3''|Cam PDE 1C|Cam-PDE 1C|hCam 3|hCam-3|HCAM3|HSPDE1C1A|Human 3'5' cyclic nucleotide Phosphodiesterase|PDE 1C|Pde1c|PDE1C_HUMAN|Phosphodiesterase 1C|Phosphodiesterase 1C calmodulin dependent|Phosphodiesterase 1C calmodulin dependent 70kDa
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14123
- Q14123
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together