Anti-PPM1F Rabbit Recombinant Polyclonal Antibody
Catalog # 77795-017
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Protein phosphatase 1F protein (1-76AA)
- Antigen Symbol:PPM1F
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P49593
- Antigen Synonyms:Ca(2+)/calmodulin-dependent protein kinase phosphatase|CaM-kinase phosphatase|CAMKP|CaMKPase|FEM 2|hFEM-2|KIAA0015|Partner of PIX 2|POPX2|PP2C phosphatase|Ppm1f|PPM1F_HUMAN|Protein fem-2 homolog|Protein phosphatase 1F (PP2C domain containing)|Protein phosphatase 1F|Protein phosphatase Mg2+/Mn2+ dependent 1F
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:5000, IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: PPM1F
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse