Anti-NPM1 Rabbit Polyclonal Antibody
Catalog # 77797-840
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from B23 around the phosphorylation site of T234.
- Antigen Symbol:NPM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P06748
- Antigen Synonyms:B23|MGC104254|NO38|NPM|NPM_HUMAN|Nucleolar phosphoprotein B23|Nucleolar protein NO38|Nucleophosmin (nucleolar phosphoprotein B23 numatrin)|Nucleophosmin|nucleophosmin nucleoplasmin family member 1|Nucleophosmin/nucleoplasmin family member 1|Numatrin|OTTHUMP00000161024|OTTHUMP00000161025|OTTHUMP00000223397|OTTHUMP00000223398
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:10000
Type:
Antigen: NPM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat