Anti-SLC12A4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: SLC12A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77820-620
- 77820-619
- Antigen Symbol:
- SLC12A4
- SLC12A4
- Antigen Name:
- Fusion protein of SLC12A4
- Fusion protein of SLC12A4
- Antigen Synonyms:
- Electroneutral potassium chloride cotransporter 1|Electroneutral potassium-chloride cotransporter 1|Erythroid K Cl cotransporter 1|Erythroid K-Cl cotransporter 1|FLJ40489|hKCC 1|hKCC1|KCC 1|KCC1|Potassium transport protein|Potassium/chloride cotransporter 1|S12A4_HUMAN|Slc12a4|Solute carrier family 12 (potassium/chloride transporters) member 4|Solute carrier family 12 member 4
- Electroneutral potassium chloride cotransporter 1|Electroneutral potassium-chloride cotransporter 1|Erythroid K Cl cotransporter 1|Erythroid K-Cl cotransporter 1|FLJ40489|hKCC 1|hKCC1|KCC 1|KCC1|Potassium transport protein|Potassium/chloride cotransporter 1|S12A4_HUMAN|Slc12a4|Solute carrier family 12 (potassium/chloride transporters) member 4|Solute carrier family 12 member 4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UP95
- Q9UP95
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together