Anti-CCDC68 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CCDC68
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77840-578
- 77840-577
- Antigen Symbol:
- CCDC68
- CCDC68
- Antigen Name:
- Coiled-coil domain-containing protein 68 protein (131-335AA)
- Coiled-coil domain-containing protein 68 protein (131-335AA)
- Antigen Synonyms:
- CCD68_HUMAN|CCDC 68|CCDC68|Coiled coil domain containing 68|Coiled coil domain containing protein 68|Coiled-coil domain-containing protein 68|CTCL associated antigen se57 1|CTCL tumor antigen se57 1|CTCL-associated antigen se57-1|Cutaneous T cell lymphoma associated antigen|Cutaneous T cell lymphoma associated antigen se57 1|Cutaneous T-cell lymphoma-associated antigen se57-1|FLJ25368|SE57 1
- CCD68_HUMAN|CCDC 68|CCDC68|Coiled coil domain containing 68|Coiled coil domain containing protein 68|Coiled-coil domain-containing protein 68|CTCL associated antigen se57 1|CTCL tumor antigen se57 1|CTCL-associated antigen se57-1|Cutaneous T cell lymphoma associated antigen|Cutaneous T cell lymphoma associated antigen se57 1|Cutaneous T-cell lymphoma-associated antigen se57-1|FLJ25368|SE57 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H2F9
- Q9H2F9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together