Anti-ZNF331 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZNF331
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77805-325
- 77805-324
- Antigen Symbol:
- ZNF331
- ZNF331
- Antigen Name:
- Zinc finger protein 331 protein (179-442AA)
- Zinc finger protein 331 protein (179-442AA)
- Antigen Synonyms:
- C2H2 like zinc finger protein|C2H2 like zinc finger protein rearranged in thyroid adenomas|C2H2-like zinc finger protein rearranged in thyroid adenomas|DKFZp686L0787|KRAB zinc finger protein|OTTHUMP00000223038|OTTHUMP00000223039|OTTHUMP00000223040|OTTHUMP00000223041|OTTHUMP00000223042|OTTHUMP00000223043|OTTHUMP00000223045|OTTHUMP00000223046|OTTHUMP00000223047|OTTHUMP00000223048|OTTHUMP00000223052|OTTHUMP00000223053|OTTHUMP00000223054|RITA|Zinc finger protein 331|Zinc finger protein 361|Zinc finger protein 463|ZN331_HUMAN|ZNF331|ZNF361|ZNF463
- C2H2 like zinc finger protein|C2H2 like zinc finger protein rearranged in thyroid adenomas|C2H2-like zinc finger protein rearranged in thyroid adenomas|DKFZp686L0787|KRAB zinc finger protein|OTTHUMP00000223038|OTTHUMP00000223039|OTTHUMP00000223040|OTTHUMP00000223041|OTTHUMP00000223042|OTTHUMP00000223043|OTTHUMP00000223045|OTTHUMP00000223046|OTTHUMP00000223047|OTTHUMP00000223048|OTTHUMP00000223052|OTTHUMP00000223053|OTTHUMP00000223054|RITA|Zinc finger protein 331|Zinc finger protein 361|Zinc finger protein 463|ZN331_HUMAN|ZNF331|ZNF361|ZNF463
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQX6
- Q9NQX6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together