Order Entry
United States
ContactUsLinkComponent
Anti-BCL2A1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-BCL2A1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-BCL2A1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77822-716
  • 77822-717
  • Antigen Symbol:
  • BCL2A1
  • BCL2A1
  • Antigen Name:
  • Bcl-2-related protein A1 protein (1-175AA)
  • Bcl-2-related protein A1 protein (1-175AA)
  • Antigen Synonyms:
  • ACC 1|ACC 2|B2LA1_HUMAN|Bcl 2 related protein A1|Bcl-2-like protein 5|Bcl-2-related protein A1|BCL2 related protein A1|Bcl2-L-5|BCL2A1|BCL2L5|BFL1|GRS|HBPA1|Hematopoietic BCL2 related protein A1|Hemopoietic specific early response protein|Hemopoietic-specific early response protein|Protein BFL 1|Protein BFL-1|Protein GRS
  • ACC 1|ACC 2|B2LA1_HUMAN|Bcl 2 related protein A1|Bcl-2-like protein 5|Bcl-2-related protein A1|BCL2 related protein A1|Bcl2-L-5|BCL2A1|BCL2L5|BFL1|GRS|HBPA1|Hematopoietic BCL2 related protein A1|Hemopoietic specific early response protein|Hemopoietic-specific early response protein|Protein BFL 1|Protein BFL-1|Protein GRS
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q16548
  • Q16548
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together