Anti-TMEM38B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: TMEM38B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77839-894
- 77839-893
- Antigen Symbol:
- TMEM38B
- TMEM38B
- Antigen Name:
- Trimeric intracellular cation channel type B protein (249-291AA)
- Trimeric intracellular cation channel type B protein (249-291AA)
- Antigen Synonyms:
- bA219P18.1|C9orf87|D4Ertd89e|FLJ10493|OTTHUMP00000021849|OTTHUMP00000063855|RP11-219P18.1|TM38B_HUMAN|TMEM38B|Transmembrane protein 38B|TRIC B|TRIC-B|TRICB|Trimeric intracellular cation channel type B
- bA219P18.1|C9orf87|D4Ertd89e|FLJ10493|OTTHUMP00000021849|OTTHUMP00000063855|RP11-219P18.1|TM38B_HUMAN|TMEM38B|Transmembrane protein 38B|TRIC B|TRIC-B|TRICB|Trimeric intracellular cation channel type B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NVV0
- Q9NVV0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together