Anti-PPP2R5C Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: PPP2R5C
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77823-893
- 77823-894
- Antigen Symbol:
- PPP2R5C
- PPP2R5C
- Antigen Name:
- Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit gamma isoform protein (1-320AA)
- Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit gamma isoform protein (1-320AA)
- Antigen Synonyms:
- 2A5G_HUMAN| B' alpha regulatory subunit| B56G| KIAA0044 | MGC23064 | PP2A B subunit B' gamma isoform| PP2A B subunit B56 gamma isoform| PP2A B subunit isoform B' gamma| PP2A B subunit isoform B''-gamma| PP2A B subunit isoform B56-gamma| PP2A B subunit isoform PR61-gamma| PP2A B subunit isoform R5-gamma| PP2A B subunit PR61 gamma isoform| PP2A B subunit R5 gamma isoform| Ppp2r5c| PR61G | Protein phosphatase 2 regulatory subunit B (B56) gamma isoform| Protein phosphatase 2 regulatory subunit B' gamma| Protein phosphatase 2| regulatory subunit B'| gamma isoform| Renal carcinoma antigen NY REN 29| Renal carcinoma antigen NY-REN-29| Serine/threonine protein phosphatase 2A 56 kDa regulatory subunit gamma isoform | Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit gamma isoform
- 2A5G_HUMAN| B' alpha regulatory subunit| B56G| KIAA0044 | MGC23064 | PP2A B subunit B' gamma isoform| PP2A B subunit B56 gamma isoform| PP2A B subunit isoform B' gamma| PP2A B subunit isoform B''-gamma| PP2A B subunit isoform B56-gamma| PP2A B subunit isoform PR61-gamma| PP2A B subunit isoform R5-gamma| PP2A B subunit PR61 gamma isoform| PP2A B subunit R5 gamma isoform| Ppp2r5c| PR61G | Protein phosphatase 2 regulatory subunit B (B56) gamma isoform| Protein phosphatase 2 regulatory subunit B' gamma| Protein phosphatase 2| regulatory subunit B'| gamma isoform| Renal carcinoma antigen NY REN 29| Renal carcinoma antigen NY-REN-29| Serine/threonine protein phosphatase 2A 56 kDa regulatory subunit gamma isoform | Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit gamma isoform
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13362
- Q13362
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together