Anti-SEMG1 Rabbit Polyclonal Antibody
Catalog # 77799-034
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Svs5
- Antigen Symbol:SEMG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:P04279
- Antigen Synonyms:Alpha-Inhibin-31|Alpha-Inhibin-92|Cancer/testis antigen 103|CT103|dJ172H20.2 (semenogelin I)|dJ172H20.2|MGC14719|Semen coagulating protein|Semenogelin|Semenogelin I|SEMG|SEMG1|SEMG1_HUMAN|Seminal basic protein|seminal vesicle secretory protein 5|SgI|Svp-1|Svp5|SVPIIA|Svs2|SVS2P|Svs2p2|Svs5
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SEMG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human