Anti-ZNF18 Rabbit Polyclonal Antibody
77821-448
New Product
- Antigen Name:Synthesized peptide derived from internal of ZNF18
- Antigen Symbol:ZNF18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P17022
- Antigen Synonyms:HDSG1|Heart development specific gene 1 protein|Heart development specific protein|Heart development-specific gene 1 protein|KOX11|Zfp 535|Zfp535|Zinc finger protein 18 (KOX 11)|Zinc finger protein 18|Zinc finger protein 535|Zinc finger protein KOX11|Zinc finger protein with KRAB and SCAN domains 6|ZKSCAN6|ZNF 535|ZNF18_HUMAN|ZNF535
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: ZNF18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat