Anti-XPNPEP1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: XPNPEP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-776
- 77839-777
- Antigen Symbol:
- XPNPEP1
- XPNPEP1
- Antigen Name:
- Xaa-Pro aminopeptidase 1 protein (2-623AA)
- Xaa-Pro aminopeptidase 1 protein (2-623AA)
- Antigen Synonyms:
- Aminoacylproline aminopeptidase|aminopeptidase P, cytosolic|APP1|Cytosolic aminopeptidase P|RP11 451M19.1|sAmp|Soluble aminopeptidase P|soluble|X Pro aminopeptidase 1|X prolyl aminopeptidase (aminopeptidase P) 1|X prolyl aminopeptidase (Aminopeptidase P) 1 soluble|X prolyl aminopeptidase 1|X prolyl aminopeptidase 1 soluble|X-Pro aminopeptidase 1|X-prolyl aminopeptidase 1|Xaa Pro aminopeptidase 1|Xaa-Pro aminopeptidase 1|XPNPEP 1|XPNPEP|xpnpep1|XPNPEPL|XPNPEPL1|XPP1_HUMAN
- Aminoacylproline aminopeptidase|aminopeptidase P, cytosolic|APP1|Cytosolic aminopeptidase P|RP11 451M19.1|sAmp|Soluble aminopeptidase P|soluble|X Pro aminopeptidase 1|X prolyl aminopeptidase (aminopeptidase P) 1|X prolyl aminopeptidase (Aminopeptidase P) 1 soluble|X prolyl aminopeptidase 1|X prolyl aminopeptidase 1 soluble|X-Pro aminopeptidase 1|X-prolyl aminopeptidase 1|Xaa Pro aminopeptidase 1|Xaa-Pro aminopeptidase 1|XPNPEP 1|XPNPEP|xpnpep1|XPNPEPL|XPNPEPL1|XPP1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQW7
- Q9NQW7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together